Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTTAAGCCTGGCTAGAGAAGC[A/G]GCAAAGTCTATGGCCTTTCCAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC10A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC10A5 - solute carrier family 10 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFAND1 - zinc finger AN1-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170796.1 | 590 | Silent Mutation | GCC,GCT | A,A 189 | NP_001164267.1 | |
NM_001170797.1 | 590 | Silent Mutation | GCC,GCT | A,A 196 | NP_001164268.1 | |
NM_024699.2 | 590 | Silent Mutation | GCC,GCT | A,A 196 | NP_078975.2 |