Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCCGTTTCAGGAAATGCCTCTTT[C/G]ATTTTGGGAACTGGGAAGATTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDCA2 - cell division cycle associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317906.1 | 618 | Missense Mutation | TTC,TTG | F,L 10 | NP_001304835.1 | |
NM_001317907.1 | 618 | Missense Mutation | TTC,TTG | F,L 25 | NP_001304836.1 | |
NM_152562.3 | 618 | Missense Mutation | TTC,TTG | F,L 25 | NP_689775.2 | |
XM_006716294.2 | 618 | Missense Mutation | TTC,TTG | F,L 25 | XP_006716357.1 | |
XM_011544415.1 | 618 | Missense Mutation | TTC,TTG | F,L 25 | XP_011542717.1 | |
XM_011544416.2 | 618 | Missense Mutation | TTC,TTG | F,L 25 | XP_011542718.1 | |
XM_011544417.2 | 618 | Missense Mutation | TTC,TTG | F,L 25 | XP_011542719.1 | |
XM_011544418.1 | 618 | Missense Mutation | TTC,TTG | F,L 25 | XP_011542720.1 |
KCTD9 - potassium channel tetramerization domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |