Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGTAGAGGCCATCCCCCTGGGA[C/T]GAAGCAAGGGCCAGCATAGTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUGGC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUGGC - nuclear GTPase, germinal center associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010906.1 | 1185 | Silent Mutation | TCA,TCG | S,S 737 | NP_001010906.1 | |
XM_011544523.2 | 1185 | Intron | XP_011542825.1 | |||
XM_011544524.2 | 1185 | Intron | XP_011542826.1 | |||
XM_011544525.1 | 1185 | Silent Mutation | TCA,TCG | S,S 350 | XP_011542827.1 | |
XM_011544526.2 | 1185 | Intron | XP_011542828.1 | |||
XM_017013403.1 | 1185 | Intron | XP_016868892.1 |