Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCACCTGCAGGCAGAGGCCATGT[A/G]TTTGGAAAGCCAGAGGAACCAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610312 | ||||||||||||||||||||
Literature Links: |
LOC100507071 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100507071 - uncharacterized LOC100507071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIWIL2 - piwi like RNA-mediated gene silencing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135721.1 | 279 | Silent Mutation | GTA,GTG | V,V 50 | NP_001129193.1 | |
NM_018068.3 | 279 | Silent Mutation | GTA,GTG | V,V 50 | NP_060538.2 | |
XM_005273550.4 | 279 | Silent Mutation | GTA,GTG | V,V 50 | XP_005273607.1 | |
XM_005273551.3 | 279 | Silent Mutation | GTA,GTG | V,V 50 | XP_005273608.1 |