Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGAACATCCGACAGCCTGACG[C/T]CCCCAGTGCCAAGAGAAGAAAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 112264 MIM: 608511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMP1 - bone morphogenetic protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHYHIP - phytanoyl-CoA 2-hydroxylase interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099335.1 | 2517 | UTR 3 | NP_001092805.1 | |||
NM_014759.3 | 2517 | UTR 3 | NP_055574.3 | |||
XM_006716416.1 | 2517 | Intron | XP_006716479.1 | |||
XM_017014102.1 | 2517 | UTR 3 | XP_016869591.1 |