Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGTCATTCGTAATGAAATATACT[A/G]AACAGAATTTTATGATGTCAGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
XKR9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
XKR9 - XK related 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011720.1 | 530 | Missense Mutation | AAA,GAA | K,E 5 | NP_001011720.1 | |
NM_001287258.1 | 530 | UTR 5 | NP_001274187.1 | |||
NM_001287259.1 | 530 | Missense Mutation | AAA,GAA | K,E 5 | NP_001274188.1 | |
NM_001287260.1 | 530 | Missense Mutation | AAA,GAA | K,E 5 | NP_001274189.1 | |
XM_006716447.3 | 530 | Intron | XP_006716510.1 | |||
XM_011517524.2 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_011515826.1 | |
XM_011517525.2 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_011515827.1 | |
XM_011517526.2 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_011515828.1 | |
XM_011517527.2 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_011515829.1 | |
XM_017013404.1 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_016868893.1 | |
XM_017013405.1 | 530 | Missense Mutation | AAA,GAA | K,E 5 | XP_016868894.1 |