Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGTTTTATTTCCCAATAATCT[C/T]CATGAGTTTCCTGTTGCTGTGAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 122560 MIM: 606469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRH - corticotropin releasing hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000756.3 | 1279 | Missense Mutation | AAG,GAG | K,E 192 | NP_000747.1 | |
XM_017013090.1 | 1279 | Missense Mutation | AAG,GAG | K,E 192 | XP_016868579.1 | |
XM_017013091.1 | 1279 | Missense Mutation | AAG,GAG | K,E 192 | XP_016868580.1 | |
XM_017013092.1 | 1279 | Missense Mutation | AAG,GAG | K,E 192 | XP_016868581.1 | |
XM_017013093.1 | 1279 | Missense Mutation | AAG,GAG | K,E 192 | XP_016868582.1 |
TRIM55 - tripartite motif containing 55 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |