Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAATGAGGAAAAATATCACATTC[C/T]TCCTTTTTATAGTCATTGTAATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZMAT4 - zinc finger matrin-type 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135731.1 | Intron | NP_001129203.1 | ||||
NM_024645.2 | Intron | NP_078921.1 | ||||
XM_017013836.1 | Intron | XP_016869325.1 | ||||
XM_017013837.1 | Intron | XP_016869326.1 | ||||
XM_017013838.1 | Intron | XP_016869327.1 | ||||
XM_017013839.1 | Intron | XP_016869328.1 | ||||
XM_017013840.1 | Intron | XP_016869329.1 |