Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAAAAATGGCCATCTCATTGG[A/T]TTGGAATGGTAAATGTTCTCAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM2 - ADAM metallopeptidase domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278113.1 | 2001 | Missense Mutation | AAA,AAT | K,N 664 | NP_001265042.1 | |
NM_001278114.1 | 2001 | Missense Mutation | AAA,AAT | K,N 620 | NP_001265043.1 | |
NM_001464.4 | 2001 | Missense Mutation | AAA,AAT | K,N 683 | NP_001455.3 | |
XM_005273468.1 | 2001 | Missense Mutation | AAA,AAT | K,N 653 | XP_005273525.1 | |
XM_011544479.1 | 2001 | Missense Mutation | AAA,AAT | K,N 557 | XP_011542781.1 | |
XM_011544480.1 | 2001 | Missense Mutation | AAA,AAT | K,N 527 | XP_011542782.1 |