Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGCATTTGAAGGGCTTCTCCCC[A/G]GAGTGCTGCCTGATGTGTGTGCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611781 | ||||||||||||||||||||
Literature Links: |
PRDM14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRDM14 - PR domain 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024504.3 | 1738 | Silent Mutation | TCC,TCT | S,S 512 | NP_078780.1 | |
XM_011517572.1 | 1738 | Intron | XP_011515874.1 |