Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACATGTCGTGGAGGTTCCCAA[A/G]TGTCACTATCATCTGTTTCTTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604764 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZHX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZHX1 - zinc fingers and homeoboxes 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017926.2 | 3275 | Missense Mutation | ACT,ATT | T,I 857 | NP_001017926.1 | |
NM_007222.4 | 3275 | Missense Mutation | ACT,ATT | T,I 857 | NP_009153.3 |
ZHX1-C8orf76 - ZHX1-C8orf76 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204180.1 | 3275 | Intron | NP_001191109.1 |