Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTGGGCTGCCAGGAGCTTCAG[A/C]GAGTGCACCTCGAAGAGCACGCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615563 MIM: 604819 | ||||||||||||||||||||
Literature Links: |
MIR6845 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6845 - microRNA 6845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP2 - nuclear receptor binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178564.3 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | NP_848659.2 | |
XM_017013378.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868867.1 | |
XM_017013379.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868868.1 | |
XM_017013380.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868869.1 | |
XM_017013381.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868870.1 | |
XM_017013382.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868871.1 | |
XM_017013383.1 | 1073 | Silent Mutation | TCG,TCT | S,S 311 | XP_016868872.1 |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |