Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTTTATGTCAGAGCCATGTTTTC[C/T]TAGAGCCCAGGGAAATAATTGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606229 MIM: 607268 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGO2 - argonaute 2, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHRAC1 - chromatin accessibility complex 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017444.5 | Intron | NP_059140.1 |