Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCTGGCTTTATTCAGCCCCAC[A/G]TGTCCAAGCATTGTCTGGGATAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 171885 | ||||||||||||||||||||
Literature Links: |
MTFR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTFR1 - mitochondrial fission regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145838.1 | 1404 | Intron | NP_001139310.1 | |||
NM_001145839.1 | 1404 | Intron | NP_001139311.1 | |||
NM_014637.3 | 1404 | Intron | NP_055452.3 | |||
XM_006716484.2 | 1404 | Intron | XP_006716547.2 | |||
XM_011517626.1 | 1404 | Intron | XP_011515928.1 | |||
XM_011517627.2 | 1404 | Intron | XP_011515929.1 | |||
XM_011517628.2 | 1404 | Intron | XP_011515930.1 |
PDE7A - phosphodiesterase 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242318.2 | 1404 | Silent Mutation | CAC,CAT | H,H 443 | NP_001229247.1 | |
NM_002603.3 | 1404 | Silent Mutation | CAC,CAT | H,H 417 | NP_002594.1 | |
XM_011517540.2 | 1404 | Silent Mutation | CAC,CAT | H,H 417 | XP_011515842.1 | |
XM_017013538.1 | 1404 | Silent Mutation | CAC,CAT | H,H 409 | XP_016869027.1 |