Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCTCCACAGGCAGATGTTGCTA[C/T]TGGACTGACAAGTGGAACCTACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608113 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGCZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGCZ - sarcoglycan zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322879.1 | 1574 | Missense Mutation | AAT,AGT | N,S 271 | NP_001309808.1 | |
NM_001322880.1 | 1574 | Missense Mutation | AAT,AGT | N,S 264 | NP_001309809.1 | |
NM_001322881.1 | 1574 | Missense Mutation | AAT,AGT | N,S 190 | NP_001309810.1 | |
NM_139167.3 | 1574 | Missense Mutation | AAT,AGT | N,S 305 | NP_631906.2 |