Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCAAGGGCCTGGCTTCACAATGA[A/G]CAGTCAGCGAATCTAAGAGCTGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614968 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2D4A - SH2 domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174159.1 | Intron | NP_001167630.1 | ||||
NM_001174160.1 | Intron | NP_001167631.1 | ||||
NM_022071.3 | Intron | NP_071354.2 | ||||
XM_011544610.1 | Intron | XP_011542912.1 | ||||
XM_017013729.1 | Intron | XP_016869218.1 |