Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTTGATCCAGGATATGCACTCAA[A/G]CTGAAATTCATGCCCACAGCCCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
XKR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XKR5 - XK related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289973.1 | 4676 | UTR 3 | NP_001276902.1 | |||
NM_207411.4 | 4676 | UTR 3 | NP_997294.3 | |||
XM_011534750.2 | 4676 | Intron | XP_011533052.1 | |||
XM_017013401.1 | 4676 | Intron | XP_016868890.1 | |||
XM_017013402.1 | 4676 | Intron | XP_016868891.1 |