Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTATGACCTGGCGGAGGAGGA[C/T]GACTCGGAGCCCCTCAACTACAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 112264 MIM: 608302 MIM: 178620 | ||||||||||||||||||||
Literature Links: |
BMP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMP1 - bone morphogenetic protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199.3 | 374 | Silent Mutation | GAC,GAT | D,D 36 | NP_001190.1 | |
NM_006129.4 | 374 | Silent Mutation | GAC,GAT | D,D 36 | NP_006120.1 | |
XM_006716386.2 | 374 | Silent Mutation | GAC,GAT | D,D 36 | XP_006716449.2 | |
XM_011544617.1 | 374 | Silent Mutation | GAC,GAT | D,D 36 | XP_011542919.1 | |
XM_017013738.1 | 374 | Silent Mutation | GAC,GAT | D,D 36 | XP_016869227.1 |
LGI3 - leucine rich repeat LGI family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SFTPC - surfactant protein C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |