Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAAAGAGACTGTCATCTTGGTT[A/G]ACTTTCCCAAGTAACAAATATAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGS1 - trimethylguanosine synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317902.1 | Intron | NP_001304831.1 | ||||
NM_024831.7 | Intron | NP_079107.6 | ||||
XM_005251328.2 | Intron | XP_005251385.1 | ||||
XM_006716485.2 | Intron | XP_006716548.1 | ||||
XM_017014004.1 | Intron | XP_016869493.1 |