Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGAGCTGCAGATGCGGACGGG[C/T]GCTGAGAACCTCTACAGGTCAGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601031 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RHPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RHPN1 - rhophilin Rho GTPase binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052924.2 | 810 | Silent Mutation | GGC,GGT | G,G 53 | NP_443156.2 | |
XM_005250772.3 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_005250829.1 | |
XM_005250773.3 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_005250830.1 | |
XM_006716502.3 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_006716565.1 | |
XM_011516817.2 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_011515119.1 | |
XM_011516818.1 | 810 | Silent Mutation | GGC,GGT | G,G 89 | XP_011515120.1 | |
XM_011516819.2 | 810 | Silent Mutation | GGC,GGT | G,G 53 | XP_011515121.1 | |
XM_011516820.2 | 810 | Silent Mutation | GGC,GGT | G,G 41 | XP_011515122.1 | |
XM_011516821.2 | 810 | Silent Mutation | GGC,GGT | G,G 4 | XP_011515123.1 | |
XM_011516822.2 | 810 | Silent Mutation | GGC,GGT | G,G 4 | XP_011515124.1 | |
XM_011516823.2 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_011515125.1 | |
XM_017013013.1 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_016868502.1 | |
XM_017013014.1 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_016868503.1 | |
XM_017013015.1 | 810 | Silent Mutation | GGC,GGT | G,G 4 | XP_016868504.1 | |
XM_017013016.1 | 810 | Silent Mutation | GGC,GGT | G,G 106 | XP_016868505.1 |
RHPN1-AS1 - RHPN1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |