Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTCTTCCTCCTCTTCTTCCCG[C/T]ATTACCTTCATTAGCTCAAGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602463 MIM: 603970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPYSL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPYSL2 - dihydropyrimidinase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNMA2 - paraneoplastic Ma antigen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007257.5 | 1763 | Missense Mutation | ATA,ATG | I,M 331 | NP_009188.1 | |
XM_011544365.2 | 1763 | Missense Mutation | ATA,ATG | I,M 331 | XP_011542667.1 |