Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGATATTTCTATCAATATGCAT[C/T]GGTGCCTGGAGTAGACTGTCTTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602064 | ||||||||||||||||||||
Literature Links: |
IMPA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IMPA1 - inositol monophosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC10A5 - solute carrier family 10 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010893.2 | 408 | Silent Mutation | CCA,CCG | P,P 136 | NP_001010893.1 |
ZFAND1 - zinc finger AN1-type containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |