Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACAGTTTCTGCCCCTGTTCAGG[A/G]CTCAGAGATATAATACAGACATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608861 MIM: 607193 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1H - ATPase H+ transporting V1 subunit H | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RGS20 - regulator of G-protein signaling 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286673.1 | 183 | Missense Mutation | ACT,GCT | T,A 31 | NP_001273602.1 | |
NM_001286674.1 | 183 | UTR 5 | NP_001273603.1 | |||
NM_001286675.1 | 183 | UTR 5 | NP_001273604.1 | |||
NM_003702.4 | 183 | Intron | NP_003693.2 | |||
NM_170587.3 | 183 | Missense Mutation | ACT,GCT | T,A 31 | NP_733466.1 | |
XM_011517622.2 | 183 | Intron | XP_011515924.1 |