Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTGGTGCTGCAGGAGCGGGTGC[A/G]CATCTTTGCCCACAAGATCTACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607352 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107984124 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC107984124 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160036.1 | 5694 | Missense Mutation | CAC,CGC | H,R 299 | NP_001153508.1 | |
NM_001160037.1 | 5694 | Missense Mutation | CAC,CGC | H,R 284 | NP_001153509.1 | |
NM_015178.2 | 5694 | Missense Mutation | CAC,CGC | H,R 277 | NP_055993.2 | |
XM_017013251.1 | 5694 | Missense Mutation | CAC,CGC | H,R 299 | XP_016868740.1 |