Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGATTGGTGCTCGTTTGGAGGCA[C/T]TGGACTACTTACAAAAATGGTATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PHF20L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHF20L1 - PHD finger protein 20-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277196.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | NP_001264125.1 | |
NM_016018.4 | 376 | Silent Mutation | CTG,TTG | L,L 22 | NP_057102.4 | |
NM_198513.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | NP_940915.1 | |
XM_005250931.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_005250988.1 | |
XM_011517075.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515377.1 | |
XM_011517076.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515378.1 | |
XM_011517077.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515379.1 | |
XM_011517078.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515380.1 | |
XM_011517079.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515381.1 | |
XM_011517080.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515382.1 | |
XM_011517081.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515383.1 | |
XM_011517088.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_011515390.1 | |
XM_017013509.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016868998.1 | |
XM_017013510.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016868999.1 | |
XM_017013511.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016869000.1 | |
XM_017013512.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016869001.1 | |
XM_017013513.1 | 376 | UTR 5 | XP_016869002.1 | |||
XM_017013514.1 | 376 | UTR 5 | XP_016869003.1 | |||
XM_017013515.1 | 376 | Intron | XP_016869004.1 | |||
XM_017013516.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016869005.1 | |
XM_017013517.1 | 376 | Silent Mutation | CTG,TTG | L,L 22 | XP_016869006.1 |