Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATCTGTTAAGTAAGGGGAAAGA[C/T]GATTTTGTTGTCTTTTGTTTCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF623 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF623 - zinc finger protein 623 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082480.2 | 91 | Intron | NP_001075949.1 | |||
NM_001261843.1 | 91 | Intron | NP_001248772.1 | |||
NM_014789.3 | 91 | Missense Mutation | ACG,ATG | T,M 1 | NP_055604.3 | |
XM_006716708.3 | 91 | Intron | XP_006716771.1 |