Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGGAGGGCGGCAGCACTGGCA[A/G]TGCTGGCAGTGACAGCAGCACCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612974 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEPTOR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEPTOR - DEP domain containing MTOR-interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283012.1 | 218 | Missense Mutation | AAT,AGT | N,S 9 | NP_001269941.1 | |
NM_022783.3 | 218 | Missense Mutation | AAT,AGT | N,S 9 | NP_073620.2 | |
XM_017013735.1 | 218 | Intron | XP_016869224.1 |