Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGGGGAATGTCTGAGGGAAAG[C/G]TTTGAGGAGTCCTGGACACCCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610635 | ||||||||||||||||||||
Literature Links: |
CTHRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTHRC1 - collagen triple helix repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256099.1 | 398 | Missense Mutation | AGC,AGG | S,R 83 | NP_001243028.1 | |
NM_138455.3 | 398 | Missense Mutation | AGC,AGG | S,R 97 | NP_612464.1 | |
XM_011516824.2 | 398 | Missense Mutation | AGC,AGG | S,R 97 | XP_011515126.1 |