Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCCTCACTTGTGGAAAGAAAC[A/G]GGCTTGTCCTTGTCGTAGCGGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614649 MIM: 609520 | ||||||||||||||||||||
Literature Links: |
RNF170 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF170 - ring finger protein 170 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
THAP1 - THAP domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018105.2 | 294 | Silent Mutation | CCC,CCT | P,P 19 | NP_060575.1 | |
NM_199003.1 | 294 | Silent Mutation | CCC,CCT | P,P 19 | NP_945354.1 |