Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAAGCCGACCATCTGCTCCGACC[A/G]GGACAACTACTGCGTGACTGTGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601384 | ||||||||||||||||||||
Literature Links: |
LOC100133669 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100133669 - uncharacterized LOC100133669 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY6E - lymphocyte antigen 6 complex, locus E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127213.1 | 290 | Missense Mutation | CAG,CGG | Q,R 44 | NP_001120685.1 | |
NM_002346.2 | 290 | Missense Mutation | CAG,CGG | Q,R 44 | NP_002337.1 |