Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAACTTCATCGCCGCGGTGGTGG[A/T]GAAGGTGGCGCCATCGGTGGTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610700 MIM: 607773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTRA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HTRA4 - HtrA serine peptidase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153692.3 | 639 | Missense Mutation | GAG,GTG | E,V 175 | NP_710159.1 | |
XM_011544431.2 | 639 | Missense Mutation | GAG,GTG | E,V 175 | XP_011542733.1 | |
XM_011544432.1 | 639 | Missense Mutation | GAG,GTG | E,V 175 | XP_011542734.1 |
PLEKHA2 - pleckstrin homology domain containing A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |