Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGACCTGGCCAGGAAGCGGGT[A/G]CCGGGCCTGGCCCAGGAGCGGTCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604782 | ||||||||||||||||||||
Literature Links: |
ASH2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASH2L - ASH2 like histone lysine methyltransferase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105214.2 | 626 | Intron | NP_001098684.1 | |||
NM_001261832.1 | 626 | Intron | NP_001248761.1 | |||
NM_001282272.1 | 626 | UTR 5 | NP_001269201.1 | |||
NM_004674.4 | 626 | Missense Mutation | ACC,GCC | T,A 14 | NP_004665.2 | |
XM_005273682.1 | 626 | Missense Mutation | ACC,GCC | T,A 14 | XP_005273739.1 | |
XM_005273683.1 | 626 | Intron | XP_005273740.1 | |||
XM_006716412.1 | 626 | Missense Mutation | ACC,GCC | T,A 14 | XP_006716475.1 | |
XM_006716413.2 | 626 | Intron | XP_006716476.1 |