Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCCATAGGATAAGCTATGTGAAT[C/G]TGGTTGTATTTTGTCCAACATTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIPAL2 - NIPA like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321635.1 | 1325 | Missense Mutation | CAT,GAT | H,D 357 | NP_001308564.1 | |
NM_001321636.1 | 1325 | Missense Mutation | ACA,AGA | T,R 325 | NP_001308565.1 | |
NM_024759.2 | 1325 | Missense Mutation | ACA,AGA | T,R 366 | NP_079035.1 | |
XM_011517302.2 | 1325 | Intron | XP_011515604.1 |