Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAACTTTGATTCTATGAAGTG[C/T]GGTATGTGTGCATGTGTGTGTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLAG1 - PLAG1 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114634.1 | 3939 | UTR 3 | NP_001108106.1 | |||
NM_001114635.1 | 3939 | UTR 3 | NP_001108107.1 | |||
NM_002655.2 | 3939 | UTR 3 | NP_002646.2 | |||
XM_011517544.2 | 3939 | UTR 3 | XP_011515846.1 | |||
XM_017013576.1 | 3939 | UTR 3 | XP_016869065.1 | |||
XM_017013577.1 | 3939 | UTR 3 | XP_016869066.1 |