Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGGGAGGTTCTTAACTAGTTAAA[C/T]AGTTGTTGGAAAAGTGCACCTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601922 MIM: 607117 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPT2 - angiopoietin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001118887.1 | 3786 | UTR 3 | NP_001112359.1 | |||
NM_001118888.1 | 3786 | UTR 3 | NP_001112360.1 | |||
NM_001147.2 | 3786 | UTR 3 | NP_001138.1 | |||
XM_017013318.1 | 3786 | UTR 3 | XP_016868807.1 |
MCPH1 - microcephalin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172574.1 | 3786 | Intron | NP_001166045.1 | |||
NM_001172575.1 | 3786 | Intron | NP_001166046.1 | |||
NM_001322042.1 | 3786 | Intron | NP_001308971.1 | |||
NM_001322043.1 | 3786 | Intron | NP_001308972.1 | |||
NM_001322045.1 | 3786 | Intron | NP_001308974.1 | |||
NM_024596.4 | 3786 | Intron | NP_078872.2 | |||
XM_011534755.2 | 3786 | Intron | XP_011533057.1 | |||
XM_011534756.2 | 3786 | Intron | XP_011533058.1 | |||
XM_011534757.2 | 3786 | Intron | XP_011533059.1 | |||
XM_011534758.2 | 3786 | Intron | XP_011533060.1 | |||
XM_011534759.2 | 3786 | Intron | XP_011533061.1 | |||
XM_011534760.2 | 3786 | Intron | XP_011533062.1 | |||
XM_017013829.1 | 3786 | Intron | XP_016869318.1 | |||
XM_017013830.1 | 3786 | Intron | XP_016869319.1 | |||
XM_017013831.1 | 3786 | Intron | XP_016869320.1 | |||
XM_017013832.1 | 3786 | Intron | XP_016869321.1 | |||
XM_017013833.1 | 3786 | Intron | XP_016869322.1 | |||
XM_017013834.1 | 3786 | Intron | XP_016869323.1 |