Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCTTGCAGCTGTCGGTGACCCT[C/G]TCCACGGTGTCTGTGTTCACTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138251 MIM: 609564 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRINA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRINA - glutamate ionotropic receptor NMDA type subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000837.1 | 697 | Silent Mutation | CTC,CTG | L,L 177 | NP_000828.1 | |
NM_001009184.1 | 697 | Silent Mutation | CTC,CTG | L,L 177 | NP_001009184.1 |
PARP10 - poly(ADP-ribose) polymerase family member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |