Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATCAGAACGGGCAAACCTCCT[C/T]TCACAACCTTTCCAGCTACAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLF10 - Kruppel like factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032282.3 | 1522 | Silent Mutation | GAA,GAG | E,E 396 | NP_001027453.1 | |
NM_005655.3 | 1522 | Silent Mutation | GAA,GAG | E,E 407 | NP_005646.1 |
LOC101927245 - vegetative cell wall protein gp1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |