Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTACGTGTCTCCAAGAAGCCC[C/T]GGATGGATGGACTTTATCCAGCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605933 | ||||||||||||||||||||
Literature Links: |
DCSTAMP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCSTAMP - dendrocyte expressed seven transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257317.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | NP_001244246.1 | |
NM_030788.3 | 151 | Silent Mutation | CCC,CCT | P,P 24 | NP_110415.1 | |
XM_011517321.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | XP_011515623.1 | |
XM_011517324.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | XP_011515626.1 | |
XM_017013878.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | XP_016869367.1 | |
XM_017013879.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | XP_016869368.1 | |
XM_017013880.1 | 151 | Silent Mutation | CCC,CCT | P,P 24 | XP_016869369.1 |