Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGTCTTCCTCCTTATGCATGA[G/T]GAACACAAGATACAGACAACTATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602155 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105379356 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105379356 - uncharacterized LOC105379356 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017014155.1 | Intron | XP_016869644.1 | ||||
XM_017014156.1 | Intron | XP_016869645.1 |
UBXN8 - UBX domain protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282189.1 | Intron | NP_001269118.1 | ||||
NM_001282199.1 | Intron | NP_001269128.1 | ||||
NM_005671.3 | Intron | NP_005662.1 | ||||
XM_011544655.2 | Intron | XP_011542957.1 | ||||
XM_017013862.1 | Intron | XP_016869351.1 |