Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGTCACAGAAGAGGTCGGCCAG[C/G]TCATCCCGCTCCTCCAGCTCGTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610085 | ||||||||||||||||||||
Literature Links: |
FAM167A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM167A - family with sequence similarity 167 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053279.2 | 1420 | Missense Mutation | GAC,GAG | D,E 176 | NP_444509.2 | |
XM_005272398.4 | 1420 | Missense Mutation | GAC,GAG | D,E 176 | XP_005272455.1 | |
XM_011543837.1 | 1420 | Missense Mutation | GAC,GAG | D,E 176 | XP_011542139.1 | |
XM_011543838.2 | 1420 | Missense Mutation | GAC,GAG | D,E 176 | XP_011542140.1 | |
XM_011543840.2 | 1420 | Missense Mutation | GAC,GAG | D,E 176 | XP_011542142.1 |
FAM167A-AS1 - FAM167A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |