Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACATTAAGAGGATGCTAGAAAA[A/G]TCAAGCTTGTTTGCTTCTGCATTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605262 MIM: 603398 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDRG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDRG1 - N-myc downstream regulated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135242.1 | 1921 | UTR 3 | NP_001128714.1 | |||
NM_001258432.1 | 1921 | UTR 3 | NP_001245361.1 | |||
NM_001258433.1 | 1921 | UTR 3 | NP_001245362.1 | |||
NM_006096.3 | 1921 | UTR 3 | NP_006087.2 | |||
XM_011516791.1 | 1921 | UTR 3 | XP_011515093.1 | |||
XM_011516792.1 | 1921 | UTR 3 | XP_011515094.1 |
WISP1 - WNT1 inducible signaling pathway protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |