Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCACTGAAGGCGTGCCCACATT[C/T]ACCACACACATAGGGCTTCTCCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF250 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF250 - zinc finger protein 250 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109689.3 | 1480 | Missense Mutation | AAA,GAA | K,E 450 | NP_001103159.1 | |
NM_021061.4 | 1480 | Missense Mutation | AAA,GAA | K,E 455 | NP_066405.1 | |
XM_005272327.1 | 1480 | Missense Mutation | AAA,GAA | K,E 450 | XP_005272384.1 | |
XM_005272328.3 | 1480 | Missense Mutation | AAA,GAA | K,E 450 | XP_005272385.1 | |
XM_006716612.3 | 1480 | Missense Mutation | AAA,GAA | K,E 455 | XP_006716675.1 | |
XM_006716613.3 | 1480 | Missense Mutation | AAA,GAA | K,E 455 | XP_006716676.1 | |
XM_011517209.1 | 1480 | Missense Mutation | AAA,GAA | K,E 450 | XP_011515511.1 |