Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACCATCTCTCCATACTCTAGA[A/C]ATTGCCTATAGAAAATAAGTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604289 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD54B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAD54B - RAD54 homolog B (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205262.2 | 2384 | Intron | NP_001192191.1 | |||
NM_001205263.1 | 2384 | Missense Mutation | ATG,ATT | M,I 567 | NP_001192192.1 | |
NM_012415.3 | 2384 | Missense Mutation | ATG,ATT | M,I 751 | NP_036547.1 |