Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATATTGTGAAACCAAAAACAAA[C/T]GCCTTTTGTGAGACCAAGCTAACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606143 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FZD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FZD3 - frizzled class receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017412.3 | 340 | UTR 5 | NP_059108.1 | |||
NM_145866.1 | 340 | UTR 5 | NP_665873.1 | |||
XM_017013841.1 | 340 | Intron | XP_016869330.1 | |||
XM_017013842.1 | 340 | UTR 5 | XP_016869331.1 | |||
XM_017013843.1 | 340 | UTR 5 | XP_016869332.1 | |||
XM_017013844.1 | 340 | UTR 5 | XP_016869333.1 |
MIR4288 - microRNA 4288 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |