Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAAAGCCCGGGTGGATCTGCTT[A/G]TCTTTGAATGCCTTGATGGTCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602419 | ||||||||||||||||||||
Literature Links: |
EGR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EGR3 - early growth response 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199880.1 | 1017 | Silent Mutation | GAC,GAT | D,D 198 | NP_001186809.1 | |
NM_001199881.1 | 1017 | Silent Mutation | GAC,GAT | D,D 182 | NP_001186810.1 | |
NM_004430.2 | 1017 | Silent Mutation | GAC,GAT | D,D 236 | NP_004421.2 | |
XM_005273425.3 | 1017 | Silent Mutation | GAC,GAT | D,D 291 | XP_005273482.1 | |
XM_005273426.3 | 1017 | Silent Mutation | GAC,GAT | D,D 198 | XP_005273483.1 | |
XM_011544429.2 | 1017 | Silent Mutation | GAC,GAT | D,D 255 | XP_011542731.1 |