Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTGAAGAGGTAGGGTGAATGGC[A/G]ACTGTGTTGTCAAAGTCTGCCTTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138300 | ||||||||||||||||||||
Literature Links: |
GSR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSR - glutathione-disulfide reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000637.3 | 1412 | Silent Mutation | GTC,GTT | V,V 508 | NP_000628.2 | |
NM_001195102.1 | 1412 | Silent Mutation | GTC,GTT | V,V 479 | NP_001182031.1 | |
NM_001195103.1 | 1412 | Silent Mutation | GTC,GTT | V,V 455 | NP_001182032.1 | |
NM_001195104.1 | 1412 | Silent Mutation | GTC,GTT | V,V 426 | NP_001182033.1 |