Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGAAGGACTCCCTGGACCCCTC[C/T]TTCACGCACGCCCTGCGCCTGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610421 | ||||||||||||||||||||
Literature Links: |
KHDRBS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KHDRBS3 - KH RNA binding domain containing, signal transduction associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006558.2 | 459 | Silent Mutation | TCC,TCT | S,S 19 | NP_006549.1 | |
XM_005250757.2 | 459 | Intron | XP_005250814.1 | |||
XM_005250758.3 | 459 | Silent Mutation | TCC,TCT | S,S 19 | XP_005250815.1 | |
XM_011516796.2 | 459 | Intron | XP_011515098.1 | |||
XM_011516797.2 | 459 | Intron | XP_011515099.1 | |||
XM_011516798.1 | 459 | Silent Mutation | TCC,TCT | S,S 19 | XP_011515100.1 | |
XM_011516799.1 | 459 | Silent Mutation | TCC,TCT | S,S 19 | XP_011515101.1 |