Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCTGATTTGAAAAGCGTGTTTG[C/T]ATTATATTGTAGAAAGCTTTGGATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0196 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0196 - KIAA0196 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NSMCE2 - NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173685.2 | Intron | NP_775956.1 | ||||
XM_005250875.2 | Intron | XP_005250932.1 | ||||
XM_005250876.4 | Intron | XP_005250933.1 | ||||
XM_011516974.2 | Intron | XP_011515276.1 | ||||
XM_011516975.1 | Intron | XP_011515277.1 | ||||
XM_017013330.1 | Intron | XP_016868819.1 | ||||
XM_017013331.1 | Intron | XP_016868820.1 | ||||
XM_017013332.1 | Intron | XP_016868821.1 | ||||
XM_017013333.1 | Intron | XP_016868822.1 | ||||
XM_017013334.1 | Intron | XP_016868823.1 |