Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGATGCCTCTACACAGCTCTCAT[C/G]CTACGTGAAGCAGGTGTCCATCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LETM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LETM2 - leucine zipper and EF-hand containing transmembrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199659.2 | 932 | Missense Mutation | TCC,TGC | S,C 210 | NP_001186588.1 | |
NM_001199660.2 | 932 | Missense Mutation | TCC,TGC | S,C 54 | NP_001186589.1 | |
NM_001286787.1 | 932 | Missense Mutation | TCC,TGC | S,C 54 | NP_001273716.1 | |
NM_001286819.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | NP_001273748.1 | |
NM_001286821.1 | 932 | Intron | NP_001273750.1 | |||
NM_144652.3 | 932 | Missense Mutation | TCC,TGC | S,C 162 | NP_653253.1 | |
XM_006716291.3 | 932 | Missense Mutation | TCC,TGC | S,C 54 | XP_006716354.1 | |
XM_011544406.1 | 932 | Missense Mutation | TCC,TGC | S,C 210 | XP_011542708.1 | |
XM_011544407.1 | 932 | Missense Mutation | TCC,TGC | S,C 210 | XP_011542709.1 | |
XM_011544408.2 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_011542710.1 | |
XM_011544409.2 | 932 | Missense Mutation | TCC,TGC | S,C 76 | XP_011542711.1 | |
XM_011544410.2 | 932 | Missense Mutation | TCC,TGC | S,C 43 | XP_011542712.1 | |
XM_017013051.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868540.1 | |
XM_017013052.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868541.1 | |
XM_017013053.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868542.1 | |
XM_017013054.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868543.1 | |
XM_017013055.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868544.1 | |
XM_017013056.1 | 932 | Missense Mutation | TCC,TGC | S,C 257 | XP_016868545.1 | |
XM_017013057.1 | 932 | Missense Mutation | TCC,TGC | S,C 209 | XP_016868546.1 |