Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCATCTTGCAGTTCTGCTGGTC[A/G]AAGGGGAAGAAGGTGACGTCGATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118502 MIM: 601212 | ||||||||||||||||||||
Literature Links: |
CHRNA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA2 - cholinergic receptor nicotinic alpha 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000742.3 | 1187 | Silent Mutation | TTC,TTT | F,F 192 | NP_000733.2 | |
NM_001282455.1 | 1187 | Silent Mutation | TTC,TTT | F,F 177 | NP_001269384.1 | |
XM_005273397.2 | 1187 | Silent Mutation | TTC,TTT | F,F 33 | XP_005273454.1 | |
XM_006716282.1 | 1187 | Silent Mutation | TTC,TTT | F,F 192 | XP_006716345.1 | |
XM_011544388.1 | 1187 | Silent Mutation | TTC,TTT | F,F 192 | XP_011542690.1 | |
XM_011544389.1 | 1187 | Intron | XP_011542691.1 | |||
XM_017013003.1 | 1187 | Silent Mutation | TTC,TTT | F,F 33 | XP_016868492.1 | |
XM_017013004.1 | 1187 | Intron | XP_016868493.1 |
PTK2B - protein tyrosine kinase 2 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |